missing translation for 'onlineSavingsMsg'
Learn More

5-Methylindole 99.0+%, TCI America™

Numéro de catalogue. M03481G
Modifier l'affichage
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
M03481G 1 g
M03485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. M03481G Fournisseur TCI America Code fournisseur M03481G
Il en reste null

Identifiants chimiques

CAS 614-96-0
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.178
MDL Number MFCD00005680
InChI Key YPKBCLZFIYBSHK-UHFFFAOYSA-N
Synonym 5-methylindole, 1h-indole, 5-methyl, 5-methylindol, indole, 5-methyl, 5-methyl indole, unii-kt52363ri5, 5-methyl-indole, indole, 2, pubchem7238
PubChem CID 11978
IUPAC Name 5-methyl-1H-indole
SMILES CC1=CC2=C(C=C1)NC=C2

Spécifications

Melting Point 60°C
Color White-Yellow
Boiling Point 100°C
Quantity 1 g
Formula Weight 131.18
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methylindole
TSCA No
Recommended Storage Refrigerator