missing translation for 'onlineSavingsMsg'
Learn More

5-Methylindole, 99%

Numéro de catalogue. AAL0689303
Modifier l'affichage
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAL0689303 1 g
AAL0689306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AAL0689303 Fournisseur Thermo Scientific Chemicals Code fournisseur L0689303
Il en reste null

CAS: 614-96-0 | C9H9N | 131.178 g/mol

5-Methylindole is used as reactant for preparation of pharmaceutically active 2-oxo-1-pyrrolidine analogues, potential anticancer immunomodulators, IL2-inducible T-cell kinase (ITK) inhibitors and CRTh2 antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methylindole is used as reactant for preparation of pharmaceutically active 2-oxo-1-pyrrolidine analogues, potential anticancer immunomodulators, IL2-inducible T-cell kinase (ITK) inhibitors and CRTh2 antagonists.

Solubility
Insoluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents and heat.

Identifiants chimiques

CAS 614-96-0
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.178
MDL Number MFCD00005680
InChI Key YPKBCLZFIYBSHK-UHFFFAOYSA-N
Synonym 5-methylindole, 1h-indole, 5-methyl, 5-methylindol, indole, 5-methyl, 5-methyl indole, unii-kt52363ri5, 5-methyl-indole, indole, 2, pubchem7238
PubChem CID 11978
IUPAC Name 5-methyl-1H-indole
SMILES CC1=CC2=C(C=C1)NC=C2

Spécifications

Melting Point 58°C to 61°C
Boiling Point 100°C (3 mmHg)
Quantity 1 g
Beilstein 2228
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 131.18
Percent Purity 99%
Chemical Name or Material 5-Methylindole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 210-400-9
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only