missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl L-Glutamate 98.0+%, TCI America™

Catalog No. M16905G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16905G 5 g
M169025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M16905G Supplier TCI America Supplier No. M16905G
Only null left

Chemical Identifiers

CAS 1499-55-4
Molecular Formula C6H11NO4
Molecular Weight (g/mol) 161.157
InChI Key ZGEYCCHDTIDZAE-BYPYZUCNSA-N
Synonym l-glutamic acid 5-methyl ester, h-glu ome-oh, polymethylglutamate, 2s-2-amino-5-methoxy-5-oxopentanoic acid, l-glutamic acid gamma-methyl ester, 5-methyl l-glutamate, l-glutamic acid, 5-methyl ester, 2s-2-amino-5-methoxy-5-oxo-pentanoic acid, 2s-2-amino-4-methoxycarbonyl butanoic acid
PubChem CID 68662
IUPAC Name (2S)-2-amino-5-methoxy-5-oxopentanoic acid
SMILES COC(=O)CCC(C(=O)O)N

Specifications

Melting Point 177°C
Color White-Yellow
Quantity 5 g
Formula Weight 161.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methyl L-Glutamate
EINECSNumber (2)-1312&(9)-0418
TSCA No