missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-3-phenylisoxazole-4-carboxaldehyde, 97%

Catalog No. AAH3212903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3212903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3212903 Supplier Thermo Scientific Chemicals Supplier No. H3212903

CAS: 87967-95-1 | C11H9NO2 | 187.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 87967-95-1
Molecular Formula C11H9NO2
Molecular Weight (g/mol) 187.20
MDL Number MFCD02677714
InChI Key SMSBKEHQYUYAHK-UHFFFAOYSA-N
Synonym 5-methyl-3-phenyl-4-isoxazolecarbaldehyde, 5-methyl-3-phenylisoxazole-4-carbaldehyde, 5-methyl-3-phenylisoxazole-4-carboxaldehyde, 5-methyl-3-phenyl-4-isoxazolecarboxaldehyde, 5-methyl-3-phenyl-isoxazole-4-carbaldehyde, acmc-20am24, 3-phenyl-5-methyl-4-isoxazolecarbaldehyde, 3-phenyl-5-methylisoxazole-4-carbaldehyde, 4-isoxazolecarboxaldehyde,5-methyl-3-phenyl, 5-methyl-3-phenyl-4-isoxazole carboxaldehyde
PubChem CID 2776144
IUPAC Name 5-methyl-3-phenyl-1,2-oxazole-4-carbaldehyde
SMILES CC1=C(C=O)C(=NO1)C1=CC=CC=C1

Specifications

Melting Point 51°C to 53°C
Quantity 1 g
Sensitivity Light and moisture sensitive
Formula Weight 187.2
Percent Purity 97%
Chemical Name or Material 5-Methyl-3-phenylisoxazole-4-carboxaldehyde
Hazard Category H302
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only