Learn More
5-Methyl-3-phenyl-4-isoxazolesulfonyl chloride, 97%, Thermo Scientific™
Supplier:  Thermo Scientific Chemicals CC00503CB
Quantity 250mg
Packaging Amber Glass Bottle
 View more versions of this product
Catalog No. CC00503CB
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Specifications
Chemical Identifiers
857283-56-8
257.688
GXQBTNFENYVPFM-UHFFFAOYSA-N
18525696
CC1=C(C(=NO1)C2=CC=CC=C2)S(=O)(=O)Cl
C10H8ClNO3S
MFCD06658966
5-methyl-3-phenyl-4-isoxazolesulfonyl chloride, 5-methyl-3-phenylisoxazole-4-sulphonyl chloride, 5-methyl-3-phenylisoxazole-4-sulfonyl chloride, chloro 5-methyl-3-phenylisoxazol-4-yl sulfone, 5-methyl-3-phenyl-isoxazole-4-sulfonyl chloride
5-methyl-3-phenyl-1,2-oxazole-4-sulfonyl chloride
Specifications
5-Methyl-3-phenyl-4-isoxazolesulfonyl chloride
96.5
97%
C10H8ClNO3S
MFCD06658966
GXQBTNFENYVPFM-UHFFFAOYSA-N
5-methyl-3-phenyl-1,2-oxazole-4-sulfonyl chloride
18525696
97%
857283-56-8
100.0
Amber Glass Bottle
250mg
5-methyl-3-phenyl-4-isoxazolesulfonyl chloride, 5-methyl-3-phenylisoxazole-4-sulphonyl chloride, 5-methyl-3-phenylisoxazole-4-sulfonyl chloride, chloro 5-methyl-3-phenylisoxazol-4-yl sulfone, 5-methyl-3-phenyl-isoxazole-4-sulfonyl chloride
CC1=C(C(=NO1)C2=CC=CC=C2)S(=O)(=O)Cl
257.688
257.7
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger