missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-3,4-diphenylisoxazole 98.0+%, TCI America™

Catalog No. M28301G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28301G 1 g
M28305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28301G Supplier TCI America Supplier No. M28301G
Only null left

Chemical Identifiers

CAS 37928-17-9
Molecular Formula C16H13NO
Molecular Weight (g/mol) 235.286
MDL Number MFCD18449672
InChI Key ZXIRUKJWLADSJS-UHFFFAOYSA-N
Synonym 5-methyl-3,4-diphenylisoxazole, 5-methyl-3,4-diphenyl-isoxazole, isoxazole, 5-methyl-3,4-diphenyl, d04ypp, 3,4-diphenyl-5-methylisoxazole, 3,4-diphenyl-5-methyl isoxazole, 3,4-diphenyl-5-methyl-isoxazole, 5-methyl-3,4-diphenyl isoxazole
PubChem CID 9991673
IUPAC Name 5-methyl-3,4-diphenyl-1,2-oxazole
SMILES CC1=C(C(=NO1)C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 98°C
Color White-Yellow
Quantity 1 g
Formula Weight 235.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methyl-3,4-diphenylisoxazole