missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-thiopheneboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. M18501G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M18501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M18501G Supplier TCI America Supplier No. M18501G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 5-Methyl-2-thiopheneboronic Acid (contains varying amounts of Anhydride)
CAS 162607-20-7
Melting Point 150°C
Color Yellow
Molecular Formula C5H7BO2S
Quantity 1 g
MDL Number MFCD01318166
Synonym 5-methylthiophene-2-boronic acid, 5-methylthiophen-2-yl boronic acid, 5-methyl-2-thiopheneboronic acid, 5-methyl-2-thienyl boronic acid, 5-methyl-2-thienylboric acid, 5-methylthiophen-2-yl boranediol, 5-methyl-2-thienylboronic acid, 5-methylthiophene-2-boronicacid, 2-methylthiophene-5-boronic acid, 2-methylthienyl-5-boronic acid
InChI Key NRIYPIBRPGAWDD-UHFFFAOYSA-N
SMILES CC1=CC=C(S1)B(O)O
IUPAC Name (5-methylthiophen-2-yl)boronic acid
Molecular Weight (g/mol) 141.98
PubChem CID 2734374
Formula Weight 141.98
Physical Form Crystalline Powder
Show More Show Less