missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-nitrobenzoic Acid 98.0+%, TCI America™

Catalog No. N04705G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N04705G 5 g
N047025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N04705G Supplier TCI America Supplier No. N04705G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3113-72-2
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00007368
InChI Key QRRSIFNWHCKMSW-UHFFFAOYSA-N
Synonym 5-methyl-2-nitrobenzoic acid, 2-nitro-5-methylbenzoic acid, benzoic acid, 5-methyl-2-nitro, 3-methyl-6-nitrobenzoic acid, 6-nitro-m-toluic acid, m-toluic acid, 6-nitro, 6-nitro-p-toluic acid, 5-methyl-2-nitrobenzoicacid, 5-methyl-2-nitro-benzoic acid, benzoic acid,5-methyl-2-nitro
PubChem CID 18371
SMILES CC1=CC(=C(C=C1)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 136°C
Color Yellow
Quantity 5 g
Formula Weight 181.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methyl-2-nitrobenzoic Acid