missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2'-deoxycytidine, MP Biomedicals™

Catalog No. ICN19888310
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19888310 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19888310 Supplier MP Biomedicals Inc Supplier No. 0219888310

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

5-Methyl-2'-deoxycytidine, White Powder

  • 5-Methyl-2'-deoxycytidine in single-stranded DNA can act in cis to signal de novo DNA methylation.
  • It has been used in epigenetics research to investigate the dynamics of DNA methylation pattern in the control of gene expression.
  • A cytidine analog and an isostere of thymidine.
  • 5-Methyl-2'-deoxycytidine is a pyrimidine nucleoside that when incorporated into single-stranded DNA can act in cis to signal de novo DNA methylation.

Chemical Identifiers

CAS 838-07-3
MDL Number MFCD00006549
InChI Key LUCHPKXVUGJYGU-XLPZGREQSA-N
PubChem CID 440055
ChEBI CHEBI:47876
IUPAC Name 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidin-2-one
SMILES CC1=CN(C(=O)N=C1N)C2CC(C(O2)CO)O

Specifications

Melting Point 217°C to 219°C (Literature)
Color White
Assay Percent Range >99.5%
Molecular Formula C10H15N5O3
Refractive Index 1.694 (lit.)
Quantity 10 mg
Synonym 5-methyl-2'-deoxycytidine, 5-methyldeoxycytidine, 2'-deoxy-5-methylcytidine, unii-b200gv71qm, cytidine, 2'-deoxy-5-methyl, 4-amino-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-5-methylpyrimidin-2 1h-one, 5-mdc, 1-2-deoxy-beta-d-ribofuranosyl-5-methylcytosine, 5-methyl-2-deoxycytidine, 4-25-00-03727 beilstein handbook reference
Solubility Information Soluble in 1 N NaOH.
Optical Rotation [α]25/D +63°, c = 1 in 1 M NaOH (lit.)
Molecular Weight (g/mol) 241.2
Formula Weight 241.2
Vapor Pressure 8.85E-12mmHg at 25°C (lit.)
Physical Form Powder
Chemical Name or Material 5-Methyl-2'-deoxycytidine
Grade 99.6 area% (HPLC)
Show More Show Less
RTECSNumber HA3860000
storageNote1 Store at Room Temperature (15-30°C).