missing translation for 'onlineSavingsMsg'
Learn More

5-méthyl-1-phényl-4-(4,4,5,5-tétraméthyle-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 97%, Thermo Scientific™

Numéro de catalogue. CC15539DA
Modifier l'affichage
Click to view available options
Quantité:
250 mg
1 g
Emballage:
Bouteille en verre ambrée
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC15539DA 1 g Bouteille en verre ambrée
CC15539CB 250 mg Bouteille en verre ambrée
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC15539DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC15539DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 849776-88-1
Formule moléculaire C16H21BN2O2
Poids moléculaire (g/mol) 284.17
Numéro MDL MFCD06797463
Clé InChI XSUARWLKVRBBKT-UHFFFAOYSA-N
Synonyme 5-methyl-1-phenyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 5-methyl-1-phenyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 5-methyl-1-phenyl-1h-pyrazole-4-boronic acid, pinacol ester, 5-methyl-1-phenyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, pubchem18509, 5-methyl-1-phenyl-1h-pyrazole-4-boronic acid pinacol ester, 5-methyl-1-phenylpyrazole-4-boronic acid pinacol ester, 5-methyl-1-phenyl-1h-pyrazol-4-yl boronic acid pinacol ester, 1h-pyrazole, 5-methyl-1-phenyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
PubChem CID 2795327
Nom de l’IUPAC 5-méthyl-1-phényl-4-(4,4,5,5-tétraméthyl-1,3,2-dioxaborolan-2-yl)pyrazole
SOURIRES CC1=C(C=NN1C1=CC=CC=C1)B1OC(C)(C)C(C)(C)O1

Spécifications

Nom ou substance chimique 5-Methyl-1-phenyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Bouteille en verre ambrée
Formule linéaire Yes
Quantité 1 g
Poids de formule 284.16
Pourcentage de pureté 97%
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.