missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-1-hexyne 99.0+%, TCI America™

Catalog No. M02715ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M02715ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M02715ML Supplier TCI America Supplier No. M02715ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2203-80-7
Molecular Formula C7H12
Molecular Weight (g/mol) 96.173
MDL Number MFCD00041610
InChI Key HKNANEMUCJGPMS-UHFFFAOYSA-N
Synonym 5-methyl-1-hexyne, 1-hexyne, 5-methyl, isoheptyne, unii-lro6ea6hk1, lro6ea6hk1, isoamylacetylene, isohept-1-yne, acmc-1cg6t, hknanemucjgpms-uhfffaoysa
PubChem CID 137490
IUPAC Name 5-methylhex-1-yne
SMILES CC(C)CCC#C

Specifications

Color Colorless
Boiling Point 91°C
Quantity 5 mL
UN Number 3295
Formula Weight 96.17
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Methyl-1-hexyne