missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxysalicylic Acid 98.0+%, TCI America™

Catalog No. M12545G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12545G 5 g
M125425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M12545G Supplier TCI America Supplier No. M12545G

Chemical Identifiers

CAS 2612-02-4
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00002459
InChI Key IZZIWIAOVZOBLF-UHFFFAOYSA-N
Synonym 5-methoxysalicylic acid, benzoic acid, 2-hydroxy-5-methoxy, 5-methoxy-2-hydroxybenzoic acid, 6-hydroxy-m-anisic acid, m-anisic acid, 6-hydroxy, 2-hydroxy-5-methoxybenzadelhyde, acid5-methoxysalicylic, 2-hydroxy-5-methoxybenzoic, 5-o-methyl gentisic acid, 2-hydroxy-5-methoxy-benzoic acid
PubChem CID 75787
IUPAC Name 2-hydroxy-5-methoxybenzoic acid
SMILES COC1=CC(=C(C=C1)O)C(=O)O

Specifications

Melting Point 144°C
Color White-Yellow
Quantity 5 g
Formula Weight 168.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methoxysalicylic Acid
EINECSNumber (3)-1647
TSCA Yes