missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxypyridine-3-boronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. M28101G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28101G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M28101G Supplier TCI America Supplier No. M28101G
Only null left

Specifications

Chemical Name or Material 5-Methoxypyridine-3-boronic Acid (contains varying amounts of Anhydride)
CAS 850991-69-4
Color White
Molecular Formula C6H8BNO3
Quantity 1 g
MDL Number MFCD03425977
Synonym 5-methoxypyridine-3-boronic acid, 5-methoxypyridin-3-yl boronic acid, 3-methoxy-5-pyridineboronic acid, 5-methoxy-3-pyridyl boronic acid, 3-borono-5-methoxypyridine, 5-methoxy-3-pyridinyl boronic acid, 3-methoxypyridine-5-boronic acid, 5-methoxy-3-pyridinylboronic acid, 5-methoxy-3-pyridineboronic acid, boronic acid,b-5-methoxy-3-pyridinyl
InChI Key ISDFOFZTZUILPE-UHFFFAOYSA-N
SMILES B(C1=CC(=CN=C1)OC)(O)O
IUPAC Name (5-methoxypyridin-3-yl)boronic acid
Molecular Weight (g/mol) 152.944
PubChem CID 2763087
Formula Weight 152.94
Physical Form Crystalline Powder
Show More Show Less