missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxy-2-nitrobenzoic acid, 97%

Catalog No. AAL1334614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1334614 25 g
AAL1334606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1334614 Supplier Thermo Scientific Chemicals Supplier No. L1334614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1882-69-5 | C8H6NO5 | 196.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1882-69-5
Molecular Formula C8H6NO5
Molecular Weight (g/mol) 196.14
MDL Number MFCD00151836
InChI Key URADKXVAIGMTEG-UHFFFAOYSA-M
Synonym 5-methoxy-2-nitrobenzoic acid, 2-nitro-5-methoxybenzoic acid, 6-nitro-m-anisic acid, 5-methoxy-2-nitro-benzoic acid, benzoic acid, 5-methoxy-2-nitro, pubchem4229, acmc-1brd6, 2-nitro-5-methoxybenzoate, 2-nitro-5-methoxybenzoicacid, 5-methoxy-2-nitrobenzoicacid
PubChem CID 339209
IUPAC Name 5-methoxy-2-nitrobenzoate
SMILES COC1=CC=C(C(=C1)C([O-])=O)[N+]([O-])=O

Specifications

Melting Point 130°C to 135°C
Quantity 25 g
Beilstein 1877657
Formula Weight 197.15
Percent Purity 97%
Chemical Name or Material 5-Methoxy-2-nitrobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only