missing translation for 'onlineSavingsMsg'
Learn More

5-Mercapto-1-phenyl-1H-tetrazole 98.0+%, TCI America™

Catalog No. P0640500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0640500G 500 g
P064025G 25 g
P0640100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P0640500G Supplier TCI America Supplier No. P0640500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 86-93-1
Molecular Formula C7H6N4S
Molecular Weight (g/mol) 178.21
MDL Number MFCD00003129
InChI Key GGZHVNZHFYCSEV-UHFFFAOYSA-N
Synonym 1-phenyl-1h-tetrazole-5-thiol, 1-phenyltetrazole-5-thiol, 1-phenyl-5-mercaptotetrazole, 5-mercapto-1-phenyltetrazole, 1-phenyltetrazoline-5-thione, mercaptophenyltetrazole, phenylmercaptotetrazole, 1-phenyl-5-tetrazolethione, 5-mercapto-1-phenyltetrazol, 1-phenyltetrazole-thiol
PubChem CID 690730
ChEBI CHEBI:79916
IUPAC Name 1-phenyl-2,5-dihydro-1H-1,2,3,4-tetrazole-5-thione
SMILES S=C1N=NNN1C1=CC=CC=C1

Specifications

Melting Point 145°C
Color White
Quantity 500 g
UN Number 1325
Formula Weight 178.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Mercapto-1-phenyl-1H-tetrazole
EINECSNumber (5)-0626
RTECSNumber XF7700000
TSCA Yes