missing translation for 'onlineSavingsMsg'
Learn More

5-(Isocyanatomethyl)-1,3-benzodioxole, ≥97%, Thermo Scientific™

Numéro de catalogue. CC50806CB
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC50806CB 250mg
CC50806DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC50806CB Fournisseur Thermo Scientific Chemicals Code fournisseur CC50806CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 71217-46-4
Formule moléculaire C9H7NO3
Poids moléculaire (g/mol) 177.159
Numéro MDL MFCD08435909
Clé InChI RIUNOJGBBOBVDE-UHFFFAOYSA-N
Synonyme 5-isocyanatomethyl-2h-1,3-benzodioxole, 5-isocyanatomethyl-1,3-benzodioxole, 1,3-benzodioxole,5-isocyanatomethyl, 1,3-benzodioxole, 5-isocyanatomethyl, 3,4-methylenedioxy benzyl isocyanate, 1,3-benzodioxol-5-ylmethyl isocyanate, 5-isocyanatomethyl benzo d 1,3 dioxole, 2h-benzo 3,4-d 1,3-dioxolan-5-ylmethanisocyanate
PubChem CID 15932354
Nom de l’IUPAC 5-(isocyanatomethyl)-1,3-benzodioxole
SOURIRES C1OC2=C(O1)C=C(C=C2)CN=C=O

Spécifications

Nom ou substance chimique 5-(Isocyanatomethyl)-1, 3-benzodioxole
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Quantité 250mg
Poids de formule 177.16
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.