missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-2'-deoxyuridine, ∼99%, MP Biomedicals™

Catalog No. ICN10035701
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10035701 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10035701 Supplier MP Biomedicals Inc Supplier No. 0210035701

missing translation for 'validMainframeMsg'

5-Iodo-2'-deoxyuridine (5-IUdR; IdUrd) is a pyrimidine analog (halogenated thymidine) that has antiviral activity agains vaccinia virus (orthopoxviruses); IdUrd is used as a radiosensitizer in human cancer studies

Chemical Identifiers

CAS 54-42-2
Molecular Formula C9H11IN2O5
Molecular Weight (g/mol) 354.10
MDL Number MFCD00134656
InChI Key XQFRJNBWHJMXHO-RRKCRQDMSA-N
Synonym idoxuridine, 5-iodo-2'-deoxyuridine, idoxuridin, 5-iododeoxyuridine, iododeoxyridine, iodoxuridine, iudr, joddeoxiuridin, idoxene, allergan 211
PubChem CID 5905
ChEBI CHEBI:147675
IUPAC Name 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(I)C(=O)NC1=O

Specifications

Color White
Assay Percent Range ∼99%
Quantity 1 g
Formula Weight 354.1
Physical Form Powder
Chemical Name or Material 5-Iodo-2'-Deoxyuridine, For HPLC analysis
EINECSNumber 200-207-8