missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-2,4-dimethoxypyrimidine 98.0+%, TCI America™

Catalog No. I05311G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05311G 1 g
I05315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I05311G Supplier TCI America Supplier No. I05311G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 52522-99-3
Molecular Formula C6H7IN2O2
Molecular Weight (g/mol) 266.038
MDL Number MFCD00090865
InChI Key KNTMOGOYRYYUIE-UHFFFAOYSA-N
Synonym 2,4-dimethoxy-5-iodopyrimidine, 5-iodo-2,4-dimethoxy-pyrimidine, pyrimidine, 5-iodo-2,4-dimethoxy, pubchem6914, acmc-1apit, 5-iodo-2,6-dimethoxypyrimidine, 2,4-dimetthoxy-5-iodopyrimidine, 5-iodo-2,4-dimethoxypyrimidine, 2,4-dimethoxy-5-iodo-pyrimidine, 5-iodo-2,4-dimethoxy-pyrimi-dine
PubChem CID 263416
IUPAC Name 5-iodo-2,4-dimethoxypyrimidine
SMILES COC1=NC(=NC=C1I)OC

Specifications

Melting Point 72°C
Color Yellow
Quantity 1 g
Formula Weight 266.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Iodo-2,4-dimethoxypyrimidine