missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-1-methyl-1H-pyrazole, 97%, Thermo Scientific™

Numéro de catalogue. p-3629709
Modifier l'affichage
Click to view available options
Quantité:
1g
5g
Emballage:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC62311DA 1g Amber Glass Bottle
CC62311DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC62311DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC62311DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 34091-51-5
Formule moléculaire C4H5IN2
Poids moléculaire (g/mol) 208.00
Numéro MDL MFCD11109327
Clé InChI RJYWUQWCLZYCTI-UHFFFAOYSA-N
Synonyme 5-iodo-1-methyl-1h-pyrazole, 1-methyl-5-iodopyrazole, 1h-pyrazole,5-iodo-1-methyl, 1h-pyrazole, 5-iodo-1-methyl, 1-methyl-5-iodo-1h-pyrazole
PubChem CID 13334065
Nom de l’IUPAC 5-iodo-1-methyl-1H-pyrazole
SOURIRES CN1N=CC=C1I

Spécifications

Nom ou substance chimique 5-Iodo-1-methyl-1H-pyrazole
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber Glass Bottle
Quantité 1g
Poids de formule 208
Pourcentage de pureté 97%
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.