missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxynorbornane 2,6-Lactone 98.0+%, TCI America™

Catalog No. H14755G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14755G 5 g
H147525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14755G Supplier TCI America Supplier No. H14755G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 92343-46-9
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.165
MDL Number MFCD00988184
InChI Key SCDJKGVPVLZLED-UHFFFAOYSA-N
Synonym 6-Hydroxyhexahydro-2H-3,5-methanocyclopenta[b]furan-2-one
PubChem CID 432639
SMILES C1C2CC3C1C(C2O)OC3=O

Specifications

Melting Point 158°C
Color Yellow
Quantity 5 g
Formula Weight 154.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Hydroxynorbornane 2,6-Lactone
EINECSNumber (5)-6966
TSCA No