missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxyflavone 98.0+%, TCI America™

Catalog No. H12381G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H12381G Supplier TCI America Supplier No. H12381G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 491-78-1
Molecular Formula C15H10O3
Molecular Weight (g/mol) 238.24
MDL Number MFCD00016944
InChI Key IYBLVRRCNVHZQJ-UHFFFAOYSA-N
Synonym 5-hydroxyflavone, primuletin, 5-hydroxy-2-phenyl-4h-chromen-4-one, 5-hydroxy-2-phenyl-chromen-4-one, chembl16807, 4h-1-benzopyran-4-one,5-hydroxy-2-phenyl, primuliten, 5-hydroxy-flavone, 5-hydroxy-2-phenyl-4-benzopyrone, 5-hydroxy-2-phenyl-4h-chromen-4-on
PubChem CID 68112
IUPAC Name 5-hydroxy-2-phenyl-4H-chromen-4-one
SMILES OC1=C2C(=O)C=C(OC2=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 159°C
Color Yellow
Quantity 1 g
Formula Weight 238.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Hydroxyflavone