missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Hydroxy-1-tetralone 99.0+%, TCI America™
Supplier: TCI America H061425G
Specifications
| 5-Hydroxy-1-tetralone | |
| 208°C | |
| C10H10O2 | |
| 25 g | |
| YPPZCRZRQHFRBH-UHFFFAOYSA-N | |
| 5-hydroxy-3,4-dihydro-2H-naphthalen-1-one | |
| 119921 | |
| ≥99.0% (GC) |
| 28315-93-7 | |
| White-Yellow | |
| MFCD00001693 | |
| 5-hydroxy-1-tetralone, 1-tetralon-5-ol, 5-hydroxy-3,4-dihydronaphthalen-1 2h-one, 5-hydroy-1-tetraione, 1 2h-naphthalenone, 3,4-dihydro-5-hydroxy, 5-hydroxy-1,2,3,4-tetrahydronaphthalen-1-one, 5-hydroxytetralone, 3,4-dihydro-5-hydroxy-1 2h-naphthalenone, 1,2,3,4-tetrahydro-5-hydroxynaphthalen-1-one, 5-hydroxy-2,3,4-trihydronaphthalen-1-one | |
| C1CC2=C(C=CC=C2O)C(=O)C1 | |
| 162.188 | |
| 162.19 | |
| Crystalline Powder |
Chemical Identifiers
| 28315-93-7 | |
| 162.188 | |
| YPPZCRZRQHFRBH-UHFFFAOYSA-N | |
| 119921 | |
| C1CC2=C(C=CC=C2O)C(=O)C1 |
| C10H10O2 | |
| MFCD00001693 | |
| 5-hydroxy-1-tetralone, 1-tetralon-5-ol, 5-hydroxy-3,4-dihydronaphthalen-1 2h-one, 5-hydroy-1-tetraione, 1 2h-naphthalenone, 3,4-dihydro-5-hydroxy, 5-hydroxy-1,2,3,4-tetrahydronaphthalen-1-one, 5-hydroxytetralone, 3,4-dihydro-5-hydroxy-1 2h-naphthalenone, 1,2,3,4-tetrahydro-5-hydroxynaphthalen-1-one, 5-hydroxy-2,3,4-trihydronaphthalen-1-one | |
| 5-hydroxy-3,4-dihydro-2H-naphthalen-1-one |
Safety and Handling
TSCA : No