missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy-1-tetralone 99.0+%, TCI America™

Catalog No. p-7130635
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H06141G 1 g
H06145G 5 g
H061425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H06141G Supplier TCI America Supplier No. H06141G
Only null left

Chemical Identifiers

CAS 28315-93-7
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00001693
InChI Key YPPZCRZRQHFRBH-UHFFFAOYSA-N
Synonym 5-hydroxy-1-tetralone, 1-tetralon-5-ol, 5-hydroxy-3,4-dihydronaphthalen-1 2h-one, 5-hydroy-1-tetraione, 1 2h-naphthalenone, 3,4-dihydro-5-hydroxy, 5-hydroxy-1,2,3,4-tetrahydronaphthalen-1-one, 5-hydroxytetralone, 3,4-dihydro-5-hydroxy-1 2h-naphthalenone, 1,2,3,4-tetrahydro-5-hydroxynaphthalen-1-one, 5-hydroxy-2,3,4-trihydronaphthalen-1-one
PubChem CID 119921
IUPAC Name 5-hydroxy-3,4-dihydro-2H-naphthalen-1-one
SMILES C1CC2=C(C=CC=C2O)C(=O)C1

Specifications

Melting Point 208°C
Color White-Yellow
Quantity 1 g
Formula Weight 162.19
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Hydroxy-1-tetralone