missing translation for 'onlineSavingsMsg'
Learn More

5-Heneicosylresorcinol, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015285
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015285 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015285 Supplier Merck Emd Millipore Supplier No. 5085110MG
Only null left
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 70110-59-7
Physical Form Neat
Linear Formula C27H48O2
MDL Number MFCD09743629
Synonym 5-Heneicosyl-1,3-benzenediol; 5-Heneicosyl-1,3-dihydroxybenzene
Type Analytical Standard
Quantity 10 mg
Formula Weight 404.67
Percent Purity ≥95% (HPLC)
Grade Analytical Standard