missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco 5 ft. Nexus Horizontal Clean Bench
Nexus Horizontal Clean Benches are designed to protect non-hazardous applications by providing an ISO Class 4 clean airflow environment. The large, 5 in. touchscreen interface features Nexus OS™ software that simplifies operation, maintains records and provides user customizaton of the system.
$20,338.63 - $27,083.30
Specifications
| Product Type | Horizontal Clean Bench |
|---|---|
| Coating | Epoxy |
| Filter Type | HEPA |
| Color | Optic White |
| Height (English) | 50.1 in. |
| Catalog Number | Mfr. No. | Material | Voltage | Includes | Depth (English) Interior | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Material | Voltage | Includes | Depth (English) Interior | Price | Quantity & Availability | ||
|
17195088
|
Labconco
5350000 |
Epoxy-coated Steel | 115 V | Clean Bench Only | 21 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17194965
|
Labconco
5350010 |
Epoxy-coated Steel | 230 V | Clean Bench Only | 21 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17194943
|
Labconco
5350100 |
Stainless Steel | 115 V | Clean Bench Only | 21 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195027
|
Labconco
5350110 |
Stainless Steel | 230 V | Clean Bench Only | 21 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17194990
|
Labconco
5350200 |
Epoxy-coated Steel | 115 V | UV Lamp | 21 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195076
|
Labconco
5350210 |
Epoxy-coated Steel | 230 V | UV Lamp | 21 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195040
|
Labconco
5350300 |
Stainless Steel | 115 V | UV Lamp | 21 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195081
|
Labconco
5350310 |
Stainless Steel | 230 V | Clean Bench with UV Lamp | 21 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195092
|
Labconco
5350500 |
Epoxy-coated Steel | 115 V | Clean Bench and Base Stand | 21 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195039
|
Labconco
5350510 |
Epoxy-coated Steel | 230 V | Base Stand | 21 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195098
|
Labconco
5350600 |
Stainless Steel | 115 V | Clean Bench and Base Stand | 21 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17194959
|
Labconco
5350610 |
Stainless Steel | 230 V | Base Stand | 21 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17194953
|
Labconco
5350700 |
Epoxy-coated Steel | 115 V | UV Lamp and Base Stand | 21 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195031
|
Labconco
5350710 |
Epoxy-coated Steel | 230 V | UV Lamp and Base Stand | 21 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17194973
|
Labconco
5350800 |
Stainless Steel | 115 V | UV Lamp and Base Stand | 21 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195032
|
Labconco
5350810 |
Stainless Steel | 230 V | UV Lamp and Base Stand | 21 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17194966
|
Labconco
5351000 |
Epoxy-coated Steel | 115 V | Clean Bench Only | 26 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195099
|
Labconco
5351010 |
Epoxy-coated Steel | 230 V | Clean Bench Only | 26 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195014
|
Labconco
5351100 |
Stainless Steel | 115 V | Clean Bench Only | 26 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195078
|
Labconco
5351110 |
Stainless Steel | 230 V | Clean Bench Only | 26 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195018
|
Labconco
5351200 |
Epoxy-coated Steel | 115 V | UV Lamp | 26 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17194961
|
Labconco
5351210 |
Epoxy-coated Steel | 230 V | UV Lamp | 26 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195094
|
Labconco
5351300 |
Stainless Steel | 115 V | Clean Bench with UV Lamp | 26 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17194947
|
Labconco
5351310 |
Stainless Steel | 230 V | UV Lamp | 26 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195100
|
Labconco
5351500 |
Epoxy-coated Steel | 115 V | Base Stand | 26 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195030
|
Labconco
5351510 |
Epoxy-coated Steel | 230 V | Base Stand | 26 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17194975
|
Labconco
5351600 |
Stainless Steel | 115 V | Base Stand | 26 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17194972
|
Labconco
5351610 |
Stainless Steel | 230 V | Base Stand | 26 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17194974
|
Labconco
5351700 |
Epoxy-coated Steel | 115 V | UV Lamp and Base Stand | 26 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17194967
|
Labconco
5351710 |
Epoxy-coated Steel | 230 V | UV Lamp and Base Stand | 26 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17194992
|
Labconco
5351800 |
Stainless Steel | 115 V | UV Lamp and Base Stand | 26 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
17195049
|
Labconco
5351810 |
Stainless Steel | 230 V | UV Lamp and Base Stand | 26 in. |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- 5 ft. Nexus Horizontal Clean Bench
- ISO Class 4 laminar airflow is 10 times cleaner than the industry standard ISO Class 5
- Energy efficient ECM motor with CAP™ Technology ensures accurate airflow between certifications
Specifications
| Horizontal Clean Bench | |
| HEPA | |
| 50.1 in. | |
| 127.5 cm | |
| None |
| Epoxy | |
| Optic White | |
| 30.2 in. | |
| 76.7 cm | |
| NEMA 5-20P |