missing translation for 'onlineSavingsMsg'
Learn More

Labconco 5 ft. Nexus Horizontal Clean Bench

Catalog No. 17194975
Change view
Click to view available options
Material:
Epoxy-coated Steel
Stainless Steel
Voltage:
115 V
230 V
Includes:
Base Stand
Clean Bench and Base Stand
Clean Bench Only
Clean Bench with UV Lamp
UV Lamp
UV Lamp and Base Stand
Depth (English) Interior:
21 in.
26 in.
32 product options available for selection
Product selection table with 32 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Voltage Includes Depth (English) Interior
17194975 Stainless Steel 115 V Base Stand 26 in.
17195088 Epoxy-coated Steel 115 V Clean Bench Only 21 in.
17194965 Epoxy-coated Steel 230 V Clean Bench Only 21 in.
17194943 Stainless Steel 115 V Clean Bench Only 21 in.
17195027 Stainless Steel 230 V Clean Bench Only 21 in.
17194990 Epoxy-coated Steel 115 V UV Lamp 21 in.
17195076 Epoxy-coated Steel 230 V UV Lamp 21 in.
17195040 Stainless Steel 115 V UV Lamp 21 in.
17195081 Stainless Steel 230 V Clean Bench with UV Lamp 21 in.
17195092 Epoxy-coated Steel 115 V Clean Bench and Base Stand 21 in.
17195039 Epoxy-coated Steel 230 V Base Stand 21 in.
17195098 Stainless Steel 115 V Clean Bench and Base Stand 21 in.
17194959 Stainless Steel 230 V Base Stand 21 in.
17194953 Epoxy-coated Steel 115 V UV Lamp and Base Stand 21 in.
17195031 Epoxy-coated Steel 230 V UV Lamp and Base Stand 21 in.
17194973 Stainless Steel 115 V UV Lamp and Base Stand 21 in.
17195032 Stainless Steel 230 V UV Lamp and Base Stand 21 in.
17194966 Epoxy-coated Steel 115 V Clean Bench Only 26 in.
17195099 Epoxy-coated Steel 230 V Clean Bench Only 26 in.
17195014 Stainless Steel 115 V Clean Bench Only 26 in.
17195078 Stainless Steel 230 V Clean Bench Only 26 in.
17195018 Epoxy-coated Steel 115 V UV Lamp 26 in.
17194961 Epoxy-coated Steel 230 V UV Lamp 26 in.
17195094 Stainless Steel 115 V Clean Bench with UV Lamp 26 in.
17194947 Stainless Steel 230 V UV Lamp 26 in.
17195100 Epoxy-coated Steel 115 V Base Stand 26 in.
17195030 Epoxy-coated Steel 230 V Base Stand 26 in.
17194972 Stainless Steel 230 V Base Stand 26 in.
17194974 Epoxy-coated Steel 115 V UV Lamp and Base Stand 26 in.
17194967 Epoxy-coated Steel 230 V UV Lamp and Base Stand 26 in.
17194992 Stainless Steel 115 V UV Lamp and Base Stand 26 in.
17195049 Stainless Steel 230 V UV Lamp and Base Stand 26 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 32 options available.
32 options
Catalog No. 17194975 Supplier Labconco Supplier No. 5351600

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Nexus Horizontal Clean Benches are designed to protect non-hazardous applications by providing an ISO Class 4 clean airflow environment. The large, 5 in. touchscreen interface features Nexus OS™ software that simplifies operation, maintains records and provides user customizaton of the system.

  • 5 ft. Nexus Horizontal Clean Bench
  • ISO Class 4 laminar airflow is 10 times cleaner than the industry standard ISO Class 5
  • Energy efficient ECM motor with CAP™ Technology ensures accurate airflow between certifications

Specifications

Product Type Horizontal Clean Bench
Coating Epoxy
Filter Type HEPA
Lighting Type LED
Material Stainless Steel
Color Optic White
Depth (English) Interior 26 in.
Depth (Metric) Interior 66.6 cm
Height (English) 50.1 in.
Height (English) Interior 30.2 in.
Height (Metric) 127.5 cm
Height (Metric) Interior 76.7 cm
Includes Base Stand
IP Rating None
Plug Type NEMA 5-20P
Voltage 115 V
Width (English) 75.2 in.
Width (English) Interior 73.0 in
Width (Metric) 191.1 cm
Width (Metric) Interior 185.4 cm
Show More Show Less