missing translation for 'onlineSavingsMsg'
Learn More

5-Formyl-2-thiopheneboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. F05491G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F05491G Supplier TCI America Supplier No. F05491G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 5-Formyl-2-thiopheneboronic Acid (contains varying amounts of Anhydride)
CAS 4347-33-5
Melting Point 135°C
Molecular Formula C5H5BO3S
Quantity 1 g
MDL Number MFCD02093666
Synonym 5-formyl-2-thiopheneboronic acid, 5-formylthiophen-2-boronic acid, 5-formylthiophen-2-yl boronic acid, 5-formylthiophene-2-boronic acid, 2-formylthiophene-5-boronic acid, 5-formylthienyl-2-boronic acid, 5-formyl-2-thienyl boronic acid, boronic acid, 5-formyl-2-thienyl, 5-formyl-2-thiopheneboronicacid, pubchem1722
InChI Key DEQOVKFWRPOPQP-UHFFFAOYSA-N
SMILES OB(O)C1=CC=C(S1)C=O
IUPAC Name (5-formylthiophen-2-yl)boronic acid
Molecular Weight (g/mol) 155.96
PubChem CID 2773430
Formula Weight 155.96
Physical Form Crystalline Powder
Show More Show Less