missing translation for 'onlineSavingsMsg'
Learn More

5-Formyl-2-furanboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. F06111G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06111G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F06111G Supplier TCI America Supplier No. F06111G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 5-Formyl-2-furanboronic Acid (contains varying amounts of Anhydride)
CAS 27329-70-0
Melting Point 151°C
Molecular Formula C5H5BO4
Quantity 1 g
MDL Number MFCD01114696
Synonym 2-formylfuran-5-boronic acid, 5-formylfuran-2-yl boronic acid, 5-formyl-2-furanboronic acid, 5-formylfuran-2-boronic acid, 5-formyl-2-furylboronic acid, 5-formyl-2-furanylboronic acid, 2-formylfuran-5-boronicacid, 5-formyl-2-furyl boronic acid, 5-formyl-2-furanyl boronic acid, boronic acid, 5-formyl-2-furanyl
InChI Key JUWYQISLQJRRNT-UHFFFAOYSA-N
SMILES B(C1=CC=C(O1)C=O)(O)O
IUPAC Name (5-formylfuran-2-yl)boronic acid
Molecular Weight (g/mol) 139.901
PubChem CID 2734355
Formula Weight 139.90
Physical Form Crystalline Powder
Show More Show Less