missing translation for 'onlineSavingsMsg'
Learn More

5'-Formyl-2,2'-bithiophene-5-boronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. F05481G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05481G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F05481G Supplier TCI America Supplier No. F05481G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1369328-62-0
Molecular Formula C9H7BO3S2
Molecular Weight (g/mol) 238.082
InChI Key RDIOTSOSMWVMSX-UHFFFAOYSA-N
Synonym 5-(5-Formyl-2-thienyl)-2-thiopheneboronic Acid
PubChem CID 44629969
IUPAC Name [5-(5-formylthiophen-2-yl)thiophen-2-yl]boronic acid
SMILES B(C1=CC=C(S1)C2=CC=C(S2)C=O)(O)O

Specifications

Quantity 1 g
Formula Weight 238.08
Physical Form Crystalline Powder
Chemical Name or Material 5′-Formyl-2,2′-bithiophene-5-boronic Acid (contains varying amounts of Anhydride)
TSCA No
Recommended Storage Refrigerator