missing translation for 'onlineSavingsMsg'
Learn More

5-Fluorocytosine 98.0+%, TCI America™

Catalog No. F03211G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03211G 1 g
F03215G 5 g
F032125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F03211G Supplier TCI America Supplier No. F03211G

Chemical Identifiers

CAS 2022-85-7
Molecular Formula C4H4FN3O
Molecular Weight (g/mol) 129.09
MDL Number MFCD00006035,MFCD00179326,MFCD03547958
InChI Key XRECTZIEBJDKEO-UHFFFAOYSA-N
Synonym 5-fluorocytosine, flucytosine, ancotil, ancobon, flucytosin, 5-fluorocystosine, fluocytosine, 5-fc, 5-fluorocytosin, fluorcytosine
PubChem CID 3366
ChEBI CHEBI:5100
IUPAC Name 6-amino-5-fluoro-1,2-dihydropyrimidin-2-one
SMILES NC1=C(F)C=NC(=O)N1

Specifications

Color White
Quantity 1 g
Formula Weight 129.09
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluorocytosine
RTECSNumber HA6040000
TSCA No