missing translation for 'onlineSavingsMsg'
Learn More

5-Fluorocytosine, 99+%

Catalog No. AC258340050
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC258340050 5 g Glass bottle
AC258342500 250 mg Glass bottle
AC258340010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC258340050 Supplier Thermo Scientific Chemicals Supplier No. 258340050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2022-85-7 | C4H4FN3O | 129.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2022-85-7
Molecular Formula C4H4FN3O
Molecular Weight (g/mol) 129.09
MDL Number MFCD00006035,MFCD00179326,MFCD03547958
InChI Key XRECTZIEBJDKEO-UHFFFAOYSA-N
Synonym 5-fluorocytosine, flucytosine, ancotil, ancobon, flucytosin, 5-fluorocystosine, fluocytosine, 5-fc, 5-fluorocytosin, fluorcytosine
PubChem CID 3366
ChEBI CHEBI:5100
IUPAC Name 6-amino-5-fluoro-1H-pyrimidin-2-one
SMILES NC1=C(F)C=NC(=O)N1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 296.0°C
Color White
Loss on Drying 1.0% max. (1 g, 105°C)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 5 g
Beilstein 25, 3688
Merck Index 15, 4156
Solubility Information Solubility in water: 15g/L (25°C)
Formula Weight 129.09
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluorocytosine
Show More Show Less
EINECSNumber 217-968-7

RUO – Research Use Only