missing translation for 'onlineSavingsMsg'
Learn More

5-Fluorobenzoxazole, 97%, Thermo Scientific™

Catalog No. p-7077567
Change view
Click to view available options
Quantité:
250mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
AC455612500 250mg
AC455610010 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC455612500 Supplier Thermo Scientific Chemicals Supplier No. 455612500

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 221347-71-3
Formule moléculaire C7H4FNO
Poids moléculaire (g/mol) 137.11
Clé InChI ZRMPAEOUOPNNPZ-UHFFFAOYSA-N
Synonyme 5-fluorobenzoxazole, 5-fluorobenzo d oxazole, benzoxazole, 5-fluoro, benzoxazole,5-fluoro, pubchem21888, 5-fluoranyl-1,3-benzoxazole, benzoxazole, 5-fluoro-9ci
PubChem CID 19937167
Nom de l’IUPAC 5-fluoro-1,3-benzoxazole
SOURIRES C1=CC2=C(C=C1F)N=CO2

Specifications

Nom ou substance chimique 5-Fluorobenzoxazole
Spectre infrarouge Authentic
Fourchette de pourcentages d’essai 96% min. (GC)
Quantité 250mg
Poids de formule 137.11
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Call a POISON CENTER or doctor/physicia

RUO – Research Use Only