missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-nitrotoluene 98.0+%, TCI America™

Catalog No. F03015G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03015G 5 g
F030125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F03015G Supplier TCI America Supplier No. F03015G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 446-33-3
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.128
MDL Number MFCD00007283
InChI Key JHFOWEGCZWLHNW-UHFFFAOYSA-N
Synonym 5-fluoro-2-nitrotoluene, 3-fluoro-6-nitrotoluene, 4-fluoro-2-methylnitrobenzene, 2-nitro-5-fluoro toluene, benzene, 4-fluoro-2-methyl-1-nitro, toluene, 5-fluoro-2-nitro, 2-nitro-5-fluorotoluene, 2-methyl-4-fluoronitrobenzene, 3-methyl-4-nitrofluorobenzene, 4-fluoro-2-methyl-1-nitro-benzene
PubChem CID 94956
IUPAC Name 4-fluoro-2-methyl-1-nitrobenzene
SMILES CC1=C(C=CC(=C1)F)[N+](=O)[O-]

Specifications

Color White-Yellow
Boiling Point 98°C
Quantity 5 g
Formula Weight 155.13
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 5-Fluoro-2-nitrotoluene