missing translation for 'onlineSavingsMsg'
Learn More

5-Ethyl-2-methylpyridine 95.0+%, TCI America™

Catalog No. E0142500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0142500ML 500 mL
E014225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0142500ML Supplier TCI America Supplier No. E0142500ML

Chemical Identifiers

CAS 104-90-5
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00006344
InChI Key NTSLROIKFLNUIJ-UHFFFAOYSA-N
Synonym aldehydine, 5-ethyl-2-picoline, 2-methyl-5-ethylpyridine, aldehydkollidin, aldehydecollidine, pyridine, 5-ethyl-2-methyl, 2,5-aldehydine, 2-picoline, 5-ethyl, 3-ethyl-6-methylpyridine, 6-methyl-3-ethylpyridine
PubChem CID 7728
IUPAC Name 5-ethyl-2-methylpyridine
SMILES CCC1=CC=C(C)N=C1

Specifications

Color Yellow
Boiling Point 178°C
Quantity 500 mL
UN Number 2300
Formula Weight 121.18
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Ethyl-2-methylpyridine
EINECSNumber (5)-0714
RTECSNumber TJ6825000
TSCA Yes