missing translation for 'onlineSavingsMsg'
Learn More

5-Ethyl-2-[2-(4-nitrophenoxy)ethyl]pyridine 98.0+%, TCI America™

$40.43 - $134.75

Chemical Identifiers

CAS 85583-54-6
Molecular Formula C15H16N2O3
Molecular Weight (g/mol) 272.304
MDL Number MFCD09032965
InChI Key KGCCHRPMSPXKJE-UHFFFAOYSA-N
PubChem CID 13314806
IUPAC Name 5-ethyl-2-[2-(4-nitrophenoxy)ethyl]pyridine
SMILES CCC1=CN=C(C=C1)CCOC2=CC=C(C=C2)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
E09515G
View Documents
TCI America
E09515G
5 g
Each for $40.43
Only null left
 
E095125G
View Documents
TCI America
E095125G
25 g
Each for $134.75
Only null left
 
Description

Description

Specifications

Chemical Identifiers

85583-54-6
272.304
KGCCHRPMSPXKJE-UHFFFAOYSA-N
5-ethyl-2-[2-(4-nitrophenoxy)ethyl]pyridine
C15H16N2O3
MFCD09032965
13314806
CCC1=CN=C(C=C1)CCOC2=CC=C(C=C2)[N+](=O)[O-]

Specifications

85583-54-6
Yellow
MFCD09032965
KGCCHRPMSPXKJE-UHFFFAOYSA-N
5-ethyl-2-[2-(4-nitrophenoxy)ethyl]pyridine
13314806
≥98.0% (GC)
5-Ethyl-2-[2-(4-nitrophenoxy)ethyl]pyridine
59°C
C15H16N2O3
5 g
CCC1=CN=C(C=C1)CCOC2=CC=C(C=C2)[N+](=O)[O-]
272.304
272.30
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents