missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Ethyl-2-[2-(4-nitrophenoxy)ethyl]pyridine 98.0+%, TCI America™
Supplier: TCI America E095125G
Specifications
| 5-Ethyl-2-[2-(4-nitrophenoxy)ethyl]pyridine | |
| 59°C | |
| C15H16N2O3 | |
| 25 g | |
| CCC1=CN=C(C=C1)CCOC2=CC=C(C=C2)[N+](=O)[O-] | |
| 272.304 | |
| 272.30 | |
| Crystalline Powder |
| 85583-54-6 | |
| Yellow | |
| MFCD09032965 | |
| KGCCHRPMSPXKJE-UHFFFAOYSA-N | |
| 5-ethyl-2-[2-(4-nitrophenoxy)ethyl]pyridine | |
| 13314806 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 85583-54-6 | |
| 272.304 | |
| KGCCHRPMSPXKJE-UHFFFAOYSA-N | |
| 5-ethyl-2-[2-(4-nitrophenoxy)ethyl]pyridine |
| C15H16N2O3 | |
| MFCD09032965 | |
| 13314806 | |
| CCC1=CN=C(C=C1)CCOC2=CC=C(C=C2)[N+](=O)[O-] |
Safety and Handling
TSCA : No