accessibility menu, dialog, popup

UserName

Enjoy Up to 50% Off Spotlight Deals All Year Long

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 85583-54-6

CH3NONOO

CAS RN 85583-54-6

IUPAC Name: 5-ethyl-2-[2-(4-nitrophenoxy)ethyl]pyridine
Synonyms: 5-ethyl-2-[2-(4-nitrophenoxy)ethyl]pyridine
Molecular Weight (g/mol): 272.3
Molecular Formula: C15H16N2O3
InChi Key: KGCCHRPMSPXKJE-UHFFFAOYSA-N
SMILES: CCC1=CN=C(CCOC2=CC=C(C=C2)[N+]([O-])=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.