missing translation for 'onlineSavingsMsg'
Learn More

(5-Ethyl-1,3-dioxan-5-yl)methyl Acrylate (stabilized with MEHQ) 98.0+%, TCI America™

Catalog No. E1334100G
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E1334100G 100 g
E1334500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E1334100G Supplier TCI America Supplier No. E1334100G

Chemical Identifiers

CAS 66492-51-1
Molecular Formula C10H16O4
Molecular Weight (g/mol) 200.234
MDL Number MFCD28046653
InChI Key STGXUBIZGYMIRM-UHFFFAOYSA-N
Synonym Acrylic Acid (5-Ethyl-1,3-dioxan-5-yl)methyl Ester
PubChem CID 171789
IUPAC Name (5-ethyl-1,3-dioxan-5-yl)methyl prop-2-enoate
SMILES CCC1(COCOC1)COC(=O)C=C

Specifications

Color Yellow
Boiling Point 78°C
Quantity 100 g
UN Number 3082
Formula Weight 200.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (5-Ethyl-1,3-dioxan-5-yl)methyl Acrylate (stabilized with MEHQ)