missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Dimethylamino-2-(2-pyridylazo)phenol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 175°C |
| Color | Purple-Red |
| Quantity | 1 g |
| Formula Weight | 242.28 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 5-Dimethylamino-2-(2-pyridylazo)phenol [for Determination of Zinc in Serum] |
Safety and Handling
| TSCA | No |