missing translation for 'onlineSavingsMsg'
Learn More

5'-Deoxy-5-fluorocytidine, Thermo Scientific™

Catalog No. AC468740050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468740050 5 g
AC468740010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC468740050 Supplier Thermo Scientific Chemicals Supplier No. 468740050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Molecular Weight (g/mol) 245.21
Purity ≥94 %
Quantity 5 g
Solubility Information Miscible with methanol, DMSO
Color White to almost white