missing translation for 'onlineSavingsMsg'
Learn More

5-Cyano-2-hydroxybenzeneboronic acid, 96%

Catalog No. AAH61915MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH61915MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH61915MD Supplier Thermo Scientific Chemicals Supplier No. H61915MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1256355-57-3 | C7H6BNO3 | 162.939 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1256355-57-3
Molecular Formula C7H6BNO3
Molecular Weight (g/mol) 162.939
InChI Key QXWBHAJKEPWEBD-UHFFFAOYSA-N
Synonym 5-cyano-2-hydroxyphenyl boronic acid, acmc-209az5, boronic acid, b-5-cyano-2-hydroxyphenyl
PubChem CID 53216735
IUPAC Name (5-cyano-2-hydroxyphenyl)boronic acid
SMILES B(C1=C(C=CC(=C1)C#N)O)(O)O

Specifications

Quantity 250 mg
Formula Weight 162.94
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 5-Cyano-2-hydroxybenzeneboronic acid
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only