missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorovaleryl chloride, 96%

Catalog No. AC154771000
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC154771000 100 mL Glass bottle
AC154770250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC154771000 Supplier Thermo Scientific Chemicals Supplier No. 154771000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1575-61-7 | C5H8Cl2O | 155.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1575-61-7
Molecular Formula C5H8Cl2O
Molecular Weight (g/mol) 155.02
MDL Number MFCD00000758
InChI Key SVNNWKWHLOJLOK-UHFFFAOYSA-N
Synonym 5-chlorovaleryl chloride, valeryl chloride, 5-chloro, pentanoyl chloride, 5-chloro, 5-chlorovalerylchloride, 5-chlorovaleroyl chloride, pentanoylchloride, 5-chloro, chlorovaleryl chloride, pubchem10919, 5-chloro-pentanoylchloride, acmc-1cuj1
PubChem CID 74089
IUPAC Name 5-chloropentanoyl chloride
SMILES ClCCCCC(Cl)=O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2000g/mL
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4630 to 1.4650
Linear Formula Cl(CH2)4COCl
Quantity 100 mL
Specific Gravity 1.2
Solubility Information Solubility in water: hydrolyses
Formula Weight 155.02
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 5-Chlorovaleryl chloride, 96%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
EINECSNumber 216-403-1
RTECSNumber YV9108000

RUO – Research Use Only