missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorosalicylamide 98.0+%, TCI America™

Catalog No. C07875G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C07875G 5 g
C078725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C07875G Supplier TCI America Supplier No. C07875G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7120-43-6
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007980
InChI Key NNHMQZBVJPQCAK-UHFFFAOYSA-N
Synonym 5-chlorosalicylamide, chlorosalicylamide, benzamide, 5-chloro-2-hydroxy, 5-chlorosalicyl-amide, acmc-209ohl, 5chloro-2-hydroxy-benzamide, ksc498a5j, 5-chloro-2-hydroxy-benzamide, #, 5-chloranyl-2-oxidanyl-benzamide
PubChem CID 348094
IUPAC Name 5-chloro-2-hydroxybenzamide
SMILES NC(=O)C1=CC(Cl)=CC=C1O

Specifications

Melting Point 234°C
Quantity 5 g
Formula Weight 171.58
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chlorosalicylamide