missing translation for 'onlineSavingsMsg'
Learn More

5-(Chloromethyl)-1,3-dimethyl-1H-pyrazole, 97%, Thermo Scientific Chemicals

Catalog No. AC451560100
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC451560100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC451560100 Supplier Thermo Scientific Chemicals Supplier No. 451560100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 852227-86-2 | C6H9ClN2 | 144.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 852227-86-2
Molecular Formula C6H9ClN2
Molecular Weight (g/mol) 144.60
MDL Number MFCD07368502
InChI Key SGEZKPNUNBVVLB-UHFFFAOYSA-N
PubChem CID 4961270
IUPAC Name 5-(chloromethyl)-1,3-dimethylpyrazole
SMILES CN1N=C(C)C=C1CCl

Specifications

Melting Point 65.0°C to 66.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 10 g
Synonym 5-chloromethyl-1,3-dimethyl-1h-pyrazole, 5-chloromethyl-1,3-dimethylpyrazole, 1h-pyrazole, 5-chloromethyl-1,3-dimethyl, 5-chloromethyl-1, pubchem23625, 3-dimethyl-1h-pyrazole, 5-chloromethyl-1,3-dime..., 5-chloromethyl-1,4-dimethyl-1h-pyrazole, 1h-pyrazole,5-chloromethyl-1,3-dimethyl
Formula Weight 144.6
Percent Purity 97%
Chemical Name or Material 5-(Chloromethyl)-1,3-dimethyl-1H-pyrazole
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

RUO – Research Use Only