missing translation for 'onlineSavingsMsg'
Learn More

5-(Chloromethyl)-1,3-dimethyl-1h-pyrazole, 97%, Thermo Scientific™

Catalog No. p-4510077
Change view
Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC12524CB 250mg Amber Glass Bottle
CC12524DA 1g Amber Glass Bottle
CC12524DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC12524CB Supplier Thermo Scientific Chemicals Supplier No. CC12524CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 852227-86-2
Molecular Formula C6H9ClN2
Molecular Weight (g/mol) 144.60
MDL Number MFCD07368502
InChI Key SGEZKPNUNBVVLB-UHFFFAOYSA-N
Synonym 5-chloromethyl-1,3-dimethyl-1h-pyrazole, 5-chloromethyl-1,3-dimethylpyrazole, 1h-pyrazole, 5-chloromethyl-1,3-dimethyl, 5-chloromethyl-1, pubchem23625, 3-dimethyl-1h-pyrazole, 5-chloromethyl-1,3-dime..., 5-chloromethyl-1,4-dimethyl-1h-pyrazole, 1h-pyrazole,5-chloromethyl-1,3-dimethyl
PubChem CID 4961270
IUPAC Name 5-(chloromethyl)-1,3-dimethyl-1H-pyrazole
SMILES CN1N=C(C)C=C1CCl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 144.6
Percent Purity 97%
Chemical Name or Material 5-(Chloromethyl)-1, 3-dimethyl-1H-pyrazole
Hazard Statement GHS P Statement Causes severe skin burns and eye damage. Harmful if swallowed.
Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.