missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-o-cresol 97,0+%, TCI America™

Numéro de catalogue. C17231G
Modifier l'affichage
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
C17231G 1 g
C17235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. C17231G Fournisseur TCI America Code fournisseur C17231G

Identifiants chimiques

CAS 5306-98-9
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.58
MDL Number MFCD00060672
InChI Key KKFPXGXMSBBNJI-UHFFFAOYSA-N
PubChem CID 79192
IUPAC Name 5-chloro-2-méthylphénol
SMILES CC1=CC=C(Cl)C=C1O

Spécifications

Melting Point 74°C
Quantity 1 g
UN Number 3437
Synonym 5-chloro-o-cresol, 5-chloro-2-cresol, o-cresol, 5-chloro, phenol, 5-chloro-2-methyl, 3-chloro-6-methylphenol, 2-methyl-5-chlorophenol, 4-chloro-2-hydroxytoluene, 5-chloro-2-methyl-phenol, phenol,5-chloro-2-methyl
Formula Weight 142.58
Percent Purity ≥97.0% (GC)
Physical Form Poudre cristalline
Chemical Name or Material 5-Chloro-o-cresol
EINECSNumber (3)-0900
TSCA No