missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chloro-6-hydroxynicotinic Acid 95.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 173.55 |
| Percent Purity | ≥95.0% (GC,T) |
| Grade | Not Available |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 5-Chloro-6-hydroxynicotinic Acid |
Safety and Handling
| TSCA | No |