missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-3-hydroxypyridine, 99%

Catalog No. AAL0756814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0756814 25 g
AAL0756806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0756814 Supplier Thermo Scientific Chemicals Supplier No. L0756814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 74115-12-1 | C5H4ClNO | 129.543 g/mol

5-Chloro-3-hydroxypyridine is used in the preparation of 4-(5-chloropyridin-3-yloxy)isoquinoline

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloro-3-hydroxypyridine is used in the preparation of 4-(5-chloropyridin-3-yloxy)isoquinoline

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 74115-12-1
Molecular Formula C5H4ClNO
Molecular Weight (g/mol) 129.543
MDL Number MFCD00006379
InChI Key TUIDQYRZDZRHPQ-UHFFFAOYSA-N
Synonym 5-chloro-3-hydroxypyridine, 5-chloro-3-pyridinol, 3-chloro-5-hydroxypyridine, 3-pyridinol, 5-chloro, 5chloro-3-pyridinol, pubchem6628, 3-chloro-5-pyridinol, acmc-209otf, 5-chloranylpyridin-3-ol, 3chloro-5-hydroxypyridine
PubChem CID 582305
IUPAC Name 5-chloropyridin-3-ol
SMILES C1=C(C=NC=C1Cl)O

Specifications

Melting Point 159°C to 163°C
Quantity 25 g
Beilstein 1422550
Solubility Information Soluble in water.
Formula Weight 129.55
Percent Purity 99%
Chemical Name or Material 5-Chloro-3-hydroxypyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 277-727-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only