missing translation for 'onlineSavingsMsg'
Learn More

5'-Chloro-3-hydroxy-2',4'-dimethoxy-2-naphthanilide 98.0+%, TCI America™

Catalog No. C210125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C210125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C210125G Supplier TCI America Supplier No. C210125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 92-72-8
Molecular Formula C19H16ClNO4
Molecular Weight (g/mol) 357.79
MDL Number MFCD00021635
InChI Key XDWATWCCUTYUDE-UHFFFAOYSA-N
Synonym N-(5-Chloro-2,4-dimethoxyphenyl)-3-hydroxy-2-naphthamide, Naphthol AS-ITR, Azoic Coupling Component 12
PubChem CID 66714
IUPAC Name N-(5-chloro-2,4-dimethoxyphenyl)-3-hydroxynaphthalene-2-carboxamide
SMILES COC1=CC(=C(C=C1NC(=O)C2=CC3=CC=CC=C3C=C2O)Cl)OC

Specifications

Melting Point 200°C
Color Yellow
Quantity 25 g
Formula Weight 357.79
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 5′-Chloro-3-hydroxy-2′,4′-dimethoxy-2-naphthanilide
EINECSNumber (5)-2281
TSCA Yes