missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-thiopheneboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. C20661G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20661G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C20661G Supplier TCI America Supplier No. C20661G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 5-Chloro-2-thiopheneboronic Acid (contains varying amounts of Anhydride)
CAS 162607-18-3
Melting Point 140°C
Color White-Yellow
Molecular Formula C4H4BClO2S
Quantity 1 g
MDL Number MFCD00236030
Synonym 5-chlorothiophene-2-boronic acid, 5-chloro-2-thienylboronic acid, 5-chlorothiophen-2-yl boronic acid, 5-chloro-2-thiopheneboronic acid, 5-chlorothiophen-2-boronic acid, 5-chlorothiophen-2-yl boranediol, 5-chlorothiphene 2-boronic acid, 5-chloro-2-thienyl boronic acid, boronic acid, 5-chloro-2-thienyl, 5-chlorothiophene-2-boronicacid
InChI Key JFCLNCVCDFUJPO-UHFFFAOYSA-N
SMILES OB(O)C1=CC=C(Cl)S1
IUPAC Name (5-chlorothiophen-2-yl)boronic acid
Molecular Weight (g/mol) 162.39
PubChem CID 2734324
Formula Weight 162.39
Physical Form Crystalline Powder
Show More Show Less